missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Retinal dehydrogenase 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Retinal dehydrogenase 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8968-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16057331

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001193826.1
Retinal dehydrogenase 2 Protein
Research Use Only
ALDH1A2
Recombinant Protein
Yeast
MTSSKIEMPG EVKADPAALM ASLHLLPSPT PNLEIKYTKI FINNEWQNSE SGRVFPVYNP ATGEQVCEVQ EADKADIDKA VQAARLAFSL GSVWRRMDAS ERGRLLDKLA DLVERDRAVL ATMESLNGGK PFLQAFYVDL QGVIKTFRYY AGWADKIHGM TIPVDGDYFT FTRHEPIGVC GQIIPWNFPL LMFAWKIAPA LCCGNTVVIK PAEQTPLSAL YMGALIKEAG FPPGVINILP GYGPTAGAAI ASHIGIDKIA FTGSTEVGKL IQEAAGRSNL KRVTLELGGK SPNIIFADAD LDYAVEQAHQ GVFFNQGQCC TAGSRIFVEE SIYEEFVRRS VERAKRRVVG SPFDPTTEQG PQIDKKQYNK ILELIQSGVA EGAKLECGGK GLGRKGFFIE PTVFSNVTDD MRIAKEEIFG PVQEILRFKT MDEVIERANN SDFGLVAAVF TNDINKALTV SSAMQAGTVW INCYNALNAQ SPFGGFKMSG NGREMGEFGL REYSEVKTVT VKIPQKNS
Greater than 90% as determined by SDS-PAGE.
8854
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers