missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RGN Protein
A cDNA sequence encoding the RGN was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13305-20ug
Additional Details : Weight : 0.01000kg
Specifications
RGN Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSSIKIECVL PENCRCGESP VWEEVSNSLL FVDIPAKKVC RWDSFTKQVQ RVTMDAPVSS VALRQSGGYV ATIGTKFCAL NWKEQSAVVL ATVDNDKKNN RFNDGKVDPA GRYFAGTMAE ETAPAVLERH QGALYSLFPD HHVKKYFDQV DISNGLDWSL DHKIFYYIDS LSYSVDAFDY DLQTGQISNR RSVYKLEKEE QIPDGMCIDA EGKLWVACYN GGRVIRLDPV TGKRLQTVKL PVDKTTSCCF GGKNYSEMYV TCARDGMDPE GLLRQPEAGG IFKITGLGVK GIAPYSYAG | |
Greater than 85.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RGN protein solution (0.5 mg/ml) containing 20mM Tris-HCl, pH 8.0, 2M Urea and 20% Glycerol. |