missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Rho GDP-dissociation inhibitor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Rho GDP-dissociation inhibitor 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8308-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001172006.1 | |
Rho GDP-dissociation inhibitor 1 Protein | |
Research Use Only | |
ARHGDIA | |
Recombinant Protein | |
E. coli | |
AEQEPTAEQL AQIAAENEED EHSVNYKPPA QKSIQEIQEL DKDDESLRKY KEALLGRVAV SADPNVPNVV VTGLTLVCSS APGPLELDLT GDLESFKKQS FVLKEGVEYR IKISFRVNRE IVSGMKYIQH TYRKGVKIDK TDYMVGSYGP RAEEYEFLTP VEEAPKGMLA RGSYSIKSRF TDDDKTDHLS WEWNLTIKKD WKD | |
Greater than 90% as determined by SDS-PAGE. |
396 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |