Läs mer
enQuireBio™ Recombinant Human Rho guanine nucleotide exchange factor 18 Protein
Produktkod. 16014991
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Förpackningsstorlek:
100µg
10µg
1mg
200µg
500µg
50µg
Produktkod. 16014991
Brand: enQuireBio™ QP8503ec200ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A DNA sequence encoding the Homo sapiens (Human) Rho guanine nucleotide exchange factor 18, was expressed in the hosts and tags indicated.
Specifikationer
Tillträdesnummer NP_001124427.1
Gen-ID (Entrez) 23370
Namn Rho guanine nucleotide exchange factor 18 Protein
Kvantitet 200 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol ARHGEF18
Protein Längd Partial (see sequence information for more details).
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art E. coli
Protein Tag GST
Sekvens MTVSQKGGPQ PTPSPAGPGT QLGPITGEMD EADSAFLKFK QTADDSLSLT SPNTESIFVE DPYTASLRSE IESDGHEFEA ESWSLAVDAA YAKKQKREVV KRQDVLYELM QTEVHHVRTL KIMLKVYSRA LQEELQFSSK AIGRLFPCAD DLLETHSHFL ARLKERRQES LEEGSDRNYV IQKIGDLLVQ QFSGENGERM KEKYGVFCSG HNEAVSHYKL LLQQNKKFQN LIKKIGNFSI VRRLGVQECI LLVTQRITKY PVLVERIIQN TEAGTEDYED LTQALNLIKD IISQVDAKVS ECEKGQRLRE IAGKMDLKSS SKLKNGLTFR KEDMLQRQLH LEGMLCWKTT SGRLKDILAI LLTDVLLLLQ EKDQKYVFAS VDSKPPVISL QKL
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.