missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RNMTL1 Protein

A cDNA sequence encoding the RNMTL1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13335-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15913249

  • 46290.00 SEK / 1mg
Estimated Shipment: 15-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RNMTL1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAALVRP ARFVVRPLLQ VVQAWDLDAR RWVRALRRSP VKVVFPSGEV VEQKRAPGKQ PRKAPSEASA QEQREKQPLE ESASRAPSTW EESGLRYDKA YPGDRRLSSV MTIVKSRPFR EKQGKILLEG RRLISDALKA GAVPKMFFFS RLEYLKELPV DKLKGVSLIK VKFEDIKDWS DLVTPQGIMG IFAKPDHVKM TYPKTQLQHS LPLLLICDNL RDPGNLGTIL RSAAGAGCSK VLLTKGCVDA WEPKVLRAGM GAHFRMPIIN NLEWETVPNY LPPDTRVYVA DNCGLYAQAE MSNKASDHGW VCDQRVMKFH KYEEEEDVET GASQDWLPHV EVQSYDSDWT EAPAAVVIGG ETYGVSLESL QLAESTGGKR LLIPVVPGVD SLNSAMAASI LLFEGKRQLR GRAEDLSRDR SYH
Greater than 85.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RNMTL1 protein solution (1 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers