missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPL18A Protein
A cDNA sequence encoding the RPL18A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13342-20ug
Additional Details : Weight : 0.01000kg
Specifications
RPL18A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMKASGTL REYKVVGRCL PTPKCHTPPL YRMRIFAPNH VVAKSRFWYF VSQLKKMKKS SGEIVYCGQV FEKSPLRVKN FGIWLRYDSR SGTHNMYREY RDLTTAGAVT QCYRDMGARH RARAHSIQIM KVEEIAASKC RRPAVKQFHD SKIKFPLPHR VLRRQHKPRF TTKRPNTFF | |
Greater than 85% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RPL18A solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |