Learn More
enQuireBio™ Recombinant Human RPS20 Protein
A cDNA sequence encoding the RPS20 was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 89170.00 SEK
Specifications
Name | RPS20 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933309
|
enQuireBio™
QP13355-2UG |
2 μg |
1545.00 SEK
2µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15913309
|
enQuireBio™
QP13355-10UG |
10 μg |
2405.00 SEK
10µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923309
|
enQuireBio™
QP13355-1MG |
1 mg |
89170.00 SEK
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RPS20 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RPS20 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol, 2mM DTT and 1mM EDTA. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAFKDTG KTPVEPEVAI HRIRITLTSR NVKSLEKVCA DLIRGAKEKN LKVKGPVRMP TKTLRITTRK TPCGEGSKTW DRFQMRIHKR LIDLHSPSEI VKQITSISIE PGVELIESTD AEPMDTEGQQ YTLRSVFESP GTCPF | |
Greater than 85% as determined by SDS-PAGE. |