missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S-phase kinase-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) S-phase kinase-associated protein 2, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001230049.1 |
---|---|
Gene ID (Entrez) | 6502 |
Name | S-phase kinase-associated protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16039190
|
enQuireBio™
QP7528-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16059190
|
enQuireBio™
QP7528-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16049190
|
enQuireBio™
QP7528-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069190
|
enQuireBio™
QP7528-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079190
|
enQuireBio™
QP7528-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089190
|
enQuireBio™
QP7528-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001230049.1 | |
S-phase kinase-associated protein 1 | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
6502 | |
Research Use Only | |
SKP2 | |
Recombinant Protein | |
E. coli | |
MHRKHLQEIP DLSSNVATSF TWGWDSSKTS ELLSGMGVSA LEKEEPDSEN IPQELLSNLG HPESPPRKRL KSKGSDKDFV IVRRPKLNRE NFPGVSWDSL PDELLLGIFS CLCLPELLKV SGVCKRWYRL ASDESLWQTL DLTGKNLHPD VTGRLLSQGV IAFRCPRSFM DQPLAEHFSP FRVQHMDLSN SVIEVSTLHG ILSQCSKLQN LSLEGLRLSD PIVNTLAKNS NLVRLNLSGC SGFSEFALQT LLSSCSRLDE LNLSWCFDFT EKHVQVAVAH VSETITQLNL SGYRKNLQKS DLSTLVRRCP NLVHLDLSDS VMLKNDCFQE FFQLNYLQHL SLSRCYDIIP ETLLELGEIP TLKTLQVFGI VPDGTLQLLK EALPHLQINC SHFTTIARPT IGNKKNQEIW GIKCRLTLQK PSCL | |
Greater than 90% as determined by SDS-PAGE. |