missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100A6 Protein
A DNA sequence encoding the Homo sapiens (Human) S100A6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6640-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055439.1 | |
S100A6 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
6277 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
S100A6 | |
Recombinant Protein | |
E. coli | |
MACPLDQAIG LLVAIFHKYS GREGDKHTLS KKELKELIQK ELTIGSKLQD AEIARLMEDL DRNKDQEVNF QEYVTFLGAL ALIYNEALKG |