missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SARAF Protein

A DNA sequence encoding the Homo sapiens (Human) SARAF, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7982-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061971

  • 7265.00 SEK / 100µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001271168.1
SARAF Protein
Research Use Only
SARAF
Recombinant Protein
Yeast
SDGQYSPPPY SEYPPFSHRY QRFTNSAGPP PPGFKSEFTG PQNTGHGATS GFGSAFTGQQ GYENSGPGFW TGLGTGGILG YLFGSNRAAT PFSDSWYYPS YPPSYPGTWN RAYSPLHGGS GSYSVCSNSD TKTRTASGYG GTRRR
Greater than 90% as determined by SDS-PAGE.
51669
100 μg
Not determined.
Cytoplasmic Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers