missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SCAMP3 Protein

A DNA sequence encoding the Homo sapiens (Human) SCAMP3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8499-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064961

  • 18585.00 SEK / 1mg


Estimated Shipment
03-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_005689.2
SCAMP3 Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
10067
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SCAMP3
Recombinant Protein
E. coli
AQSRDGGNPF AEPSELDNPF QDPAVIQHRP SRQYATLDVY NPFETREPPP AYEPPAPAPL PPPSAPSLQP SRKLSPTEPK NYGSYSTQAS AAAATAELLK KQEELNRKAE ELDRRERELQ HAALGGTATR QNNWPPLPSF CPVQPCFFQD ISMEIPQEFQ KTVSTMYYL
Videos
SDS
Documents