missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SCAN domain-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) SCAN domain-containing protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6651-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064820

  • 5980.00 SEK / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_057642.1
SCAN domain-containing protein 1
Research Use Only
SCAND1
Recombinant Protein
E. coli
MAATEPILAA TGSPAAVPPE KLEGAGSSSA PERNCVGSSL PEASPPAPEP SSPNAAVPEA IPTPRAAASA ALELPLGPAP VSVAPQAEAE ARSTPGPAGS RLGPETFRQR FRQFRYQDAA GPREAFRQLR ELSRQWLRPD IRTKEQIVEM LVQEQLLAIL PEAARARRIR RRTDVRITG
Greater than 80% as determined by SDS-PAGE.
51282
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers