missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SCF / C-kit ligand Protein
A DNA sequence encoding the Homo sapiens (Human) SCF / C-kit ligand, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6264-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000890.1 | |
SCF / C-kit ligand Protein | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
4254 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Kitlg | |
Recombinant Protein | |
E. coli | |
EGICRNRVTN NVKDVTKLVA NLPKDYMITL KYVPGMDVLP SHCWISEMVV QLSDSLTDLL DKFSNISEGL SNYSIIDKLV NIVDDLVECV KENSSKDLKK SFKSPEPRLF TPEEFFRIFN RSIDAFKDFV VASETSDCVV SSTLSPEKDS RVSVTKPFML PPVA |