missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Semenogelin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Semenogelin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6662-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064900

  • 26990.00 SEK / 1mg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002998.1
Semenogelin-1 Protein
Research Use Only
SEMG1
Recombinant Protein
E. coli
QKGGSKGRLP SEFSQFPHGQ KGQHYSGQKG KQQTESKGSF SIQYTYHVDA NDHDQSRKSQ QYDLNALHKT TKSQRHLGGS QQLLHNKQEG RDHDKSKGHF HRVVIHHKGG KAHRGTQNPS QDQGNSPSGK GISSQYSNTE ERLWVHGLSK EQTSVSGAQK GRKQGGSQSS YVLQTEELVA NKQQRETKNS HQNKGHYQNV VEVREEHSSK VQTSLCPAHQ DKLQHGSKDI FSTQDELLVY NKNQHQTKNL NQDQQHGRKA NKISYQSSST EERRLHYGEN GVQKDVSQSS IYSQTEEKAQ GKSQKQITIP SQEQEHSQKA NKISYQSSST EERRLHYGEN GVQKDVSQRS IYSQTEKLVA GKSQIQAPNP KQEPWHGEN
Greater than 90% as determined by SDS-PAGE.
6406
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers