missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine protease 23 Protein
A DNA sequence encoding the Homo sapiens (Human) Serine protease 23, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6538-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001280107.1 | |
Serine protease 23 Protein | |
Research Use Only | |
PRSS23 | |
Recombinant Protein | |
E. coli | |
QVSPYSAPWK PTWPAYRLPV VLPQSTLNLA KPDFGAEAKL EVSSSCGPQC HKGTPLPTYE EAKQYLSYET LYANGSRTET QVGIYILSSS GDGAQHRDSG SSGKSRRKRQ IYGYDSRFSI FGKDFLLNYP FSTSVKLSTG CTGTLVAEKH VLTAAHCIHD GKTYVKGTQK LRVGFLKPKF KDGGRGANDS TSAMPEQMKF QWIRVKRTHV PKGWIKGNAN DIGMDYDYAL LELKKPHKRK FMKIGVSPPA KQLPGGRIHF SGYDNDRPGN LVYRFCDVKD ETYDLLYQQC DAQPGASGSG VYVRMWKRQQ QKWERKIIGI FSGHQWVDMN GSPQDFNVAV RITPLKYAQI CYWIKGNYLD CREG | |
Greater than 90% as determined by SDS-PAGE. |
11098 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |