missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sex comb on midleg-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Sex comb on midleg-like protein 4, was expressed in the hosts and tags indicated.
1580.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001273337.1 |
---|---|
Gene ID (Entrez) | 256380 |
Name | Sex comb on midleg-like protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091491
|
enQuireBio™
QP7903-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011501
|
enQuireBio™
QP7903-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001501
|
enQuireBio™
QP7903-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021501
|
enQuireBio™
QP7903-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031501
|
enQuireBio™
QP7903-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041501
|
enQuireBio™
QP7903-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001273337.1 | |
Sex comb on midleg-like protein 1 | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
256380 | |
Research Use Only | |
SCML4 | |
Recombinant Protein | |
E. coli | |
MPCQRTAWIG CDRQRTPPFH WREIKSRVLM TPLALSPPRS TPEPDLSSIP QDAATVPSLA APQALTVCLY INKQANAGPY LERKKVQQLP EHFGPERPSA VLQQAVQACI DCAHQQKLVF SLVKQGYGGE MVSVSASFDG KQHLRSLPVV NSIGYVLRFL AKLCRSLLCD DLFSHQPFPR GCSASEKVQE KEEGRMESVK TVTTEEYLVN PVGMNRYSVD TSASTFNHRG SLHPSSSLYC KRQNSGDSHL GGGPAATAGG PRTSPMSSGG PSAPGLRPPA SSPKRNTTSL EGNRCGNVMH ASASH | |
Greater than 90% as determined by SDS-PAGE. |