Läs mer
enQuireBio™ Recombinant Human SHH / Sonic hedgehog Protein
Produktkod. 15913008
Kvantitet:
100 μg
25 μg
250 μg
5 μg
Förpackningsstorlek:
100µg
250µg
25µg
5µg
Produktkod. 15913008
Brand: enQuireBio™ QP10181ec5ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A DNA sequence encoding the Human SHH / Sonic hedgehog, was expressed in the hosts and tags indicated.
Specifikationer
Tillträdesnummer NP_000184.1
Gen-ID (Entrez) 6469
Namn SHH / Sonic hedgehog Protein
Kvantitet 5 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Less than 1.0 EU/ug as determined by LAL method.
Gensymbol SHH
Protein Längd Partial (see sequence information for more details).
Biologisk aktivitet The ED50 as determined by inducing alkaline phosphatase production of murine C3H/10T1/2 cells is less than 1 ug/ml, corresponding to a specific activity of > 1.0 M 103 IU/mg.
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art E. coli
Protein Tag Untagged
Sekvens IVIGPGRGFG KRRHPKKLTP LAYKQFIPNV AEKTLGASGR YEGKISRNSE RFKELTPNYN PDIIFKDEEN TGADRLMTQR CKDKLNALAI SVMNQWPGVK LRVTEGWDED GHHSEESLHY EGRAVDITTS DRDRSKYGML ARLAVEAGFD WVYYESKAHI HCSVKAENSV AAKSGG
Buffert 0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized
Renhet eller kvalitetsklass Greater than 98% as determined by SDS-PAGE and HPLC.