missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sialidase-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Sialidase-3, was expressed in the hosts and tags indicated.
1580.00 SEK - 18390.00 SEK
Specifications
Accession Number | NP_006647.3 |
---|---|
Gene ID (Entrez) | 10825 |
Name | Sialidase-3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16047021
|
enQuireBio™
QP8892-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087021
|
enQuireBio™
QP8892-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037021
|
enQuireBio™
QP8892-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067021
|
enQuireBio™
QP8892-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077021
|
enQuireBio™
QP8892-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057021
|
enQuireBio™
QP8892-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_006647.3 | |
Sialidase-3 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
10825 | |
Research Use Only | |
NEU3 | |
Recombinant Protein | |
E. coli | |
EEVTTCSFNS PLFRQEDDRG ITYRIPALLY IPPTHTFLAF AEKRSTRRDE DALHLVLRRG LRIGQLVQWG PLKPLMEATL PGHRTMNPCP VWEQKSGCVF LFFICVRGHV TERQQIVSGR NAARLCFIYS QDAGCSWSEV RDLTEEVIGS ELKHWATFAV GPGHGIQLQS GRLVIPAYTY YIPSWFFCFQ LPCKTRPHSL MIYSDDLGVT WHHGRLIRPM VTVECEVAEV TGRAGHPVLY CSARTPNRCR AEALSTDHGE GFQRLALSRQ LCEPPHGCQG SVVSFRPLEI PHRCQDSSSK DAPTIQQSSP GSSLRLEEEA GTPSESWLLY SHPTSRKQRV DLGIYLNQTP LEAACWSRPW ILHCGPCGYS DLAALEEEGL FGCLFECGTK QECEQIAFRL FTHREILSHL QGDCTSPGRN PSQFKSN | |
Greater than 90% as determined by SDS-PAGE. |