missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SLC4A4 Protein
A cDNA sequence encoding the SLC4A4 was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 46290.00 SEK
Specifications
Name | SLC4A4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933849
|
enQuireBio™
QP13522-5UG |
5 μg |
1545.00 SEK
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923849
|
enQuireBio™
QP13522-20UG |
20 μg |
2405.00 SEK
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15913849
|
enQuireBio™
QP13522-1MG |
1 mg |
46290.00 SEK
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SLC4A4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SLC4A4 protein solution contains 20mM Tris-HCl, pH 7.5, 0.5mM DTT, 0.1mM PMSF and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMSTE NVEGKPSNLG ERGRARSSTF LRVVQPMFNH SIFTSAVSPA AERIRFILGEEDDSPAPPQL FTELDELLAV DGQEMEWKET ARWIKFEEKV EQGGERWSKP HVATLSLHSL FELRTCMEKG SIMLDREASS LPQLVEMIVDHQIETGLLKP ELKDKVTYTL LRKHRHQTKK SNLRSLADIG KTVSSASRMF TNPDNGSPAM THRNLTSSSL NDISDKPEKD QLKNKFMKKLPRDAEASNVL VGEVDFLDTP FIAFVRLQQA VMLGALTEVP VPTRFLFILL GPKGKAKSYH EIGRAIATLM SDEVFHDIAY KAKDRHDLIAGIDEFLDEVI VLPPGEWDPA IRIEPPKSLP SSDKRKNMYS GGENVQMNGD TPHDGGHGGG GHGDCEELQR TGRFCGGLIK DIKRKAPFFASDFYDALNIQ | |
Greater than 90.0% as determined by SDS-PAGE. |