missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SMARCB1 Protein
A DNA sequence encoding the Homo sapiens (Human) SMARCB1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7615-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001007469.1 | |
SMARCB1 Protein | |
Research Use Only | |
SMARCB1 | |
Recombinant Protein | |
E. coli | |
MMMMALSKTF GQKPVKFQLE DDGEFYMIGS EVGNYLRMFR GSLYKRYPSL WRRLATVEER KKIVASSHGK KTKPNTKDHG YTTLATSVTL LKASEVEEIL DGNDEKYKAV SISTEPPTYL REQKAKRNSQ WVPTLPNSSH HLDAVPCSTT INRNRMGRDK KRTFPLCFDD HDPAVIHENA SQPEVLVPIR LDMEIDGQKL RDAFTWNMNE KLMTPEMFSE ILCDDLDLNP LTFVPAIASA IRQQIESYPT DSILEDQSDQ RVIIKLNIHV GNISLVDQFE WDMSEKENSP EKFALKLCSE LGLGGEFVTT IAYSIRGQLS WHQKTYAFSE NPLPTVEIAI RNTGDADQWC PLLETLTDAE MEKKIRDQDR NTRRMRRLAN TAPAW | |
Greater than 90% as determined by SDS-PAGE. |
6598 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |