missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SMUG1 Protein
A DNA sequence encoding the Homo sapiens (Human) SMUG1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7679-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001230716.1 | |
SMUG1 Protein | |
10 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
23583 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
SMUG1 | |
Recombinant Protein | |
E. coli | |
MPQAFLLGSI HEPAGALMEP QPCPGSLAES FLEEELRLNA ELSQLQFSEP VGIIYNPVEY AWEPHRNYVT RYCQGPKEVL FLGMNPGPFG MAQTGVPFGE VSMVRDWLGI VGPVLTPPQE HPKRPVLGLE CPQSEGPRQS MGHEIKSELL MGGCSWIRGK IQCDRVQVRR PGFSSQL |