missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNCA / alpha-Synuclein Protein
A DNA sequence encoding the Homo sapiens (Human) SNCA / alpha-Synuclein, was expressed in the hosts and tags indicated.
2075.00 SEK - 21365.00 SEK
Specifications
Accession Number | NP_000336.1 |
---|---|
Gene ID (Entrez) | 6622 |
Name | SNCA / alpha-Synuclein Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16065250
|
enQuireBio™
QP6704-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005260
|
enQuireBio™
QP6704-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16055250
|
enQuireBio™
QP6704-YE-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085250
|
enQuireBio™
QP6704-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095250
|
enQuireBio™
QP6704-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075250
|
enQuireBio™
QP6704-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000336.1 | |
SNCA / alpha-Synuclein Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
6622 | |
Research Use Only | |
SNCA | |
Recombinant Protein | |
Yeast | |
MDVFMKGLSK AKEGVVAAAE KTKQGVAEAA GKTKEGVLYV GSKTKEGVVH GVATVAEKTK EQVTNVGGAV VTGVTAVAQK TVEGAGSIAA ATGFVKKDQL GKNEEGAPQE GILEDMPVDP DNEAYEMPSE EGYQDYEPEA | |
Greater than 90% as determined by SDS-PAGE. |