missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNRPC Protein
A cDNA sequence encoding the SNRPC was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13544-HIS-10ug
Additional Details : Weight : 0.01000kg
Specifications
SNRPC Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MPKFYCDYCD TYLTHDSPSV RKTHCSGRKH KENVKDYYQK WMEEQAQSLI DKTTAAFQQG KIPPTPFSAP PPAGAMIPPP PSLPGPPRPG MMPAPHMGGP PMMPMMGPPP PGMMPVGPAP GMRPPMGGHM PMMPGPPMMR PPARPMMVPT RPGMTRPDR | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SNRPC is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |