missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Solute carrier family 41 member 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Solute carrier family 41 member 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6695-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065180

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_115524.3
Solute carrier family 41 member 2 Protein
Research Use Only
SLC41A2
Recombinant Protein
E. coli
MTNSKGRSIT DKTSGGPSSG GGFVDWTLRL NTIQSDKFLN LLLSMVPVIY QKNQEDRHKK ANGIWQDGLS TAVQTFSNRS EQHMEYHSFS EQSFHANNGH ASSSCSQKYD DYANYNYCDG RETSETTAML QDEDISSDGD EDAIVEVTPK LPKESSGIMA LQ
Greater than 90% as determined by SDS-PAGE.
84102
200 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers