missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Solute carrier family 41 member 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Solute carrier family 41 member 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6695-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_115524.3 | |
Solute carrier family 41 member 2 Protein | |
Research Use Only | |
SLC41A2 | |
Recombinant Protein | |
E. coli | |
MTNSKGRSIT DKTSGGPSSG GGFVDWTLRL NTIQSDKFLN LLLSMVPVIY QKNQEDRHKK ANGIWQDGLS TAVQTFSNRS EQHMEYHSFS EQSFHANNGH ASSSCSQKYD DYANYNYCDG RETSETTAML QDEDISSDGD EDAIVEVTPK LPKESSGIMA LQ | |
Greater than 90% as determined by SDS-PAGE. |
84102 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |