missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SPG21 Protein
A cDNA sequence encoding the SPG21 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13574-20ug
Additional Details : Weight : 0.01000kg
Specifications
SPG21 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGEIKVSPDY NWFRGTVPLK KIIVDDDDSK IWSLYDAGPR SIRCPLIFLP PVSGTADVFF RQILALTGWG YRVIALQYPV YWDHLEFCDG FRKLLDHLQL DKVHLFGASL GGFLAQKFAE YTHKSPRVHS LILCNSFSDT SIFNQTWTAN SFWLMPAFML KKIVLGNFSS GPVDPMMADA IDFMVDRLES LGQSELASRL TLNCQNSYVE PHKIRDIPVT IMDVFDQSAL STEAKEEMYK LYPNARRAHL KTGGNFPYLC RSAEVNLYVQ IHLLQFHGTK YAAIDPSMVS AEELEVQKGS LGISQEEQ | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SPG21 protein solution contains 20mM Tris-HCl, pH 8. |