missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Spindlin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Spindlin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8292-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006708.2 | |
Spindlin-1 Protein | |
Research Use Only | |
SPIN1 | |
Recombinant Protein | |
E. coli | |
MKTPFGKTPG QRSRADAGHA GVSANMMKKR TSHKKHRSSV GPSKPVSQPR RNIVGCRIQH GWKEGNGPVT QWKGTVLDQV PVNPSLYLIK YDGFDCVYGL ELNKDERVSA LEVLPDRVAT SRISDAHLAD TMIGKAVEHM FETEDGSKDE WRGMVLARAP VMNTWFYITY EKDPVLYMYQ LLDDYKEGDL RIMPDSNDSP PAEREPGEVV DSLVGKQVEY AKEDGSKRTG MVIHQVEAKP SVYFIKFDDD FHIYVYDLVK TS | |
Greater than 80% as determined by SDS-PAGE. |
10927 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |