missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Spliceosome RNA helicase DDX39B Protein
A DNA sequence encoding the Homo sapiens (Human) Spliceosome RNA helicase DDX39B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8315-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004631.1 | |
Spliceosome RNA helicase DDX39B Protein | |
Research Use Only | |
DDX39B | |
Recombinant Protein | |
E. coli | |
AENDVDNELL DYEDDEVETA AGGDGAEAPA KKDVKGSYVS IHSSGFRDFL LKPELLRAIV DCGFEHPSEV QHECIPQAIL GMDVLCQAKS GMGKTAVFVL ATLQQLEPVT GQVSVLVMCH TRELAFQISK EYERFSKYMP NVKVAVFFGG LSIKKDEEVL KKNCPHIVVG TPGRILALAR NKSLNLKHIK HFILDECDKM LEQLDMRRDV QEIFRMTPHE KQVMMFSATL SKEIRPVCRK FMQDPMEIFV | |
Greater than 90% as determined by SDS-PAGE. |
7919 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |