missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Substance-P receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Substance-P receptor, was expressed in the hosts and tags indicated.
1580.00 SEK - 18390.00 SEK
Specifications
Accession Number | NP_001049.1 |
---|---|
Gene ID (Entrez) | 6869 |
Name | Substance-P receptor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16055871
|
enQuireBio™
QP8669-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095871
|
enQuireBio™
QP8669-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045871
|
enQuireBio™
QP8669-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075871
|
enQuireBio™
QP8669-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085871
|
enQuireBio™
QP8669-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065871
|
enQuireBio™
QP8669-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Spezifikation
NP_001049.1 | |
Substance-P receptor Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6869 | |
Research Use Only | |
NK1R | |
Recombinant Protein | |
E. coli | |
MDNVLPVDSD LSPNISTNTS EPNQFVQPA | |
Greater than 90% as determined by SDS-PAGE. |