missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SUCLG2 Protein

A DNA sequence encoding the Homo sapiens (Human) SUCLG2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7942-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061741

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001171070.1
SUCLG2 Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
8801
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SUCLG2
Recombinant Protein
E. coli
MSDNGVRVQR FFVADTANEA LEAAKRLNAK EIVLKAQILA GGRGKGVFNS GLKGGVHLTK DPNVVGQLAK QMIGYNLATK QTPKEGVKVN KVMVAEALDI SRETYLAILM DRSCNGPVLV GSPQGGVDIE EVAASNPELI FKEQIDIFEG IKDSQAQRMA ENLGFVGPLK SQAADQITKL YNLFLKIDAT QVEVNPFGET PEGQVVCFDA KINFDDNAEF RQKDIFAMDD KSENEPIENE AAKYDLKYIG LDGNIACFVN GAGLAMATCD IIFLNGGKPA NFLDLGGGVK EAQVYQAFKL LTADPKVEAI LVNIFGGIVN CAIIANGITK ACRELELKVP LVVRLEGTNV QEAQKILNNS GLPITSAIDL EDAAK
Videos
SDS
Documents