missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SULT1A3 Protein
A DNA sequence encoding the Homo sapiens (Human) SULT1A3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6748-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001017390.1 | |
SULT1A3 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
105369243 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
SULT1A3 | |
Recombinant Protein | |
E. coli | |
MELIQDTSRP PLEYVKGVPL IKYFAEALGP LQSFQARPDD LLINTYPKSG TTWVSQILDM IYQGGDLEKC NRAPIYVRVP FLEVNDPGEP SGLETLKDTP PPRLIKSHLP LALLPQTLLD QKVKVVYVAR NPKDVAVSYY HFHRMEKAHP EPGTWDSFLE KFMAGEVSYG SWYQHVQEWW ELSRTHPVLY LFYEDMKENP KREIQKILEF VGRSLPEETM DFMVQHTSFK EMKKNPMTNY TTVPQELMDH SISPFMRKGM AGDWKTTFTV AQNERFDADY AEKMAGCSLS FRSEL |