missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Suppressor of cytokine signaling 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Suppressor of cytokine signaling 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6711-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003736.1 | |
Suppressor of cytokine signaling 1 Protein | |
Research Use Only | |
SOCS1 | |
Recombinant Protein | |
E. coli | |
MVAHNQVAAD NAVSTAAEPR RRPEPSSSSS SSPAAPARPR PCPAVPAPAP GDTHFRTFRS HADYRRITRA SALLDACGFY WGPLSVHGAH ERLRAEPVGT FLVRDSRQRN CFFALSVKMA SGPTSIRVHF QAGRFHLDGS RESFDCLFEL LEHYVAAPRR MLGAPLRQRR VRPLQELCRQ RIVATVGREN LARIPLNPVL RDYLSSFPFQ I | |
Greater than 91.1% as determined by SDS-PAGE. |
8651 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |