missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SUPT3H Protein
A DNA sequence encoding the Homo sapiens (Human) SUPT3H, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6752-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003590.1 | |
SUPT3H Protein | |
Research Use Only | |
SUPT3H | |
Recombinant Protein | |
E. coli | |
MVVFGTTFFN FDSFIGKTDS IEIIGKSVFP YCYHNILPLL NDSGRYSLGD ARRPLHETAV LVEDVVHTQL INLLQQAAEV SQLRGARVIT PEDLLFLMRK DKKKLRRLLK YMFIRDYKSK IVKGIDEDDL LEDKLSGSNN ANKRQKIAQD FLNSIDQTGE LLAMFEDDEI DEVKQERMER AERQTRIMDS AQYAEFCESR QLSFSKKASK FRDWLDCSSM EIKPNVVAME ILAYLAYETV AQLVDLALLV RQDMVTKAGD PFSHAISATF IQYHNSAEST AACGVEAHSD AIQPCHIREA IRRYSHRIGP LSPFTNAYRR NGMAFLAC | |
Greater than 90% as determined by SDS-PAGE. |
8464 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |