missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SUPT3H Protein

A DNA sequence encoding the Homo sapiens (Human) SUPT3H, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6752-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065570

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_003590.1
SUPT3H Protein
Research Use Only
SUPT3H
Recombinant Protein
E. coli
MVVFGTTFFN FDSFIGKTDS IEIIGKSVFP YCYHNILPLL NDSGRYSLGD ARRPLHETAV LVEDVVHTQL INLLQQAAEV SQLRGARVIT PEDLLFLMRK DKKKLRRLLK YMFIRDYKSK IVKGIDEDDL LEDKLSGSNN ANKRQKIAQD FLNSIDQTGE LLAMFEDDEI DEVKQERMER AERQTRIMDS AQYAEFCESR QLSFSKKASK FRDWLDCSSM EIKPNVVAME ILAYLAYETV AQLVDLALLV RQDMVTKAGD PFSHAISATF IQYHNSAEST AACGVEAHSD AIQPCHIREA IRRYSHRIGP LSPFTNAYRR NGMAFLAC
Greater than 90% as determined by SDS-PAGE.
8464
200 μg
Not determined.
Partial (see sequence information for more details). of Isoform 3
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions