missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SUPT3H Protein

A DNA sequence encoding the Homo sapiens (Human) SUPT3H, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6752-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16055570

  • 18585.00 SEK / 1mg


Estimated Shipment
08-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_003590.1
SUPT3H Protein
1 mg
Not determined.
Partial (see sequence information for more details). of Isoform 3
Human
His-SUMO
Tris-based buffer, 50% glycerol
8464
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SUPT3H
Recombinant Protein
E. coli
MVVFGTTFFN FDSFIGKTDS IEIIGKSVFP YCYHNILPLL NDSGRYSLGD ARRPLHETAV LVEDVVHTQL INLLQQAAEV SQLRGARVIT PEDLLFLMRK DKKKLRRLLK YMFIRDYKSK IVKGIDEDDL LEDKLSGSNN ANKRQKIAQD FLNSIDQTGE LLAMFEDDEI DEVKQERMER AERQTRIMDS AQYAEFCESR QLSFSKKASK FRDWLDCSSM EIKPNVVAME ILAYLAYETV AQLVDLALLV RQDMVTKAGD PFSHAISATF IQYHNSAEST AACGVEAHSD AIQPCHIREA IRRYSHRIGP LSPFTNAYRR NGMAFLAC
Videos
SDS
Documents