missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SURF2 Protein
A cDNA sequence encoding the SURF2 was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 46290.00 SEK
Specifications
Name | SURF2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934279
|
enQuireBio™
QP13652-5UG |
5 μg |
1545.00 SEK
5µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924279
|
enQuireBio™
QP13652-20UG |
20 μg |
2405.00 SEK
20µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15914279
|
enQuireBio™
QP13652-1MG |
1 mg |
46290.00 SEK
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SURF2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SURF2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol and 2mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSELPGD VRAFLREHPS LRLQTDARKV RCILTGHELP CRLPELQVYT RGKKYQRLVR ASPAFDYAEF EPHIVPSTKN PHQLFCKLTL RHINKCPEHV LRHTQGRRYQ RALCKYEECQ KQGVEYVPAC LVHRRRRRED QMDGDGPRPR EAFWEPTSSD EGGAASDDSM TDLYPPELFT RKDLGSTEDG DGTDDFLTDK EDEKAKPPRE KATDEGRRET TVYRGLVQKR GKKQLGSLKK KFKSHHRKPK SFSSCKQPG | |
Greater than 90.0% as determined by SDS-PAGE. |