missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Synaptotagmin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Synaptotagmin-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6757-ec-500ug
Dettagli aggiuntivi : Weight : 0.01000kg
Specifica
NP_001129277.1 | |
Synaptotagmin-1 Protein | |
Research Use Only | |
SYT1 | |
Recombinant Protein | |
E. coli | |
KNAINMKDVK DLGKTMKDQA LKDDDAETGL TDGEEKEEPK EEEKLGKLQY SLDYDFQNNQ LLVGIIQAAE LPALDMGGTS DPYVKVFLLP DKKKKFETKV HRKTLNPVFN EQFTFKVPYS ELGGKTLVMA VYDFDRFSKH DIIGEFKVPM NTVDFGHVTE EWRDLQSAEK EEQEKLGDIC FSLRYVPTAG KLTVVILEAK NLKKMDVGGL SDPYVKIHLM QNGKRLKKKK TTIKKNTLNP YYNESFSFEV PFEQIQKVQV VVTVLDYDKI GKNDAIGKVF VGYNSTGAEL RHWSDMLANP RRPIAQWHTL QVEEEVDA | |
Greater than 90% as determined by SDS-PAGE. |
6857 | |
500 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |