missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TAF12 Protein
A DNA sequence encoding the Homo sapiens (Human) TAF12, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7622-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001128690.1 | |
TAF12 Protein | |
Research Use Only | |
TAF12 | |
Recombinant Protein | |
E. coli | |
MNQFGPSALI NLSNFSSIKP EPASTPPQGS MANSTAVVKI PGTPGAGGRL SPENNQVLTK KKLQDLVREV DPNEQLDEDV EEMLLQIADD FIESVVTAAC QLARHRKSST LEVKDVQLHL ERQWNMWIPG FGSEEIRPYK KACTTEAHKQ RMALIRKTTK K | |
Greater than 90% as determined by SDS-PAGE. |
6883 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |