missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Talin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Talin-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8291-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055874.2 | |
Talin-2 Protein | |
Research Use Only | |
TLN2 | |
Recombinant Protein | |
E. coli | |
RPQKIRMLDG SVKTVMVDDS KTVGELLVTI CSRIGITNYE EYSLIQETIE EKKEEGTGTL KKDRTLLRDE RKMEKLKAKL HTDDDLNWLD HSRTFREQGV DENETLLLRR KFFYSDQNVD SRDPVQLNLL YVQARDDILN GSHPVSFEKA CEFGGFQAQI QFGPHVEHKH KPGFLDLKEF LPKEYIKQRG AEKRIFQEHK NCGEMSEIEA KVKYVKLARS LRTYGVSFFL VKEKMKGKNK LVPRLLGITK DSVMRVDEKT KEVLQEWPLT TVKRWAASPK SFTLDFGEYQ ESYYSVQTTE GEQISQLIAG YIDIILKKK | |
Greater than 90% as determined by SDS-PAGE. |
83660 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |