missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TAR DNA-binding protein 43 Protein

A DNA sequence encoding the Homo sapiens (Human) TAR DNA-binding protein 43, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6761-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015640

  • 22640.00 SEK / 1mg
Estimated Shipment: 14-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_031401.1
TAR DNA-binding protein 43 Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
23435
Greater than 80% as determined by SDS-PAGE.
Research Use Only
TARDBP
Recombinant Protein
E. coli
MSEYIRVTED ENDEPIEIPS EDDGTVLLST VTAQFPGACG LRYRNPVSQC MRGVRLVEGI LHAPDAGWGN LVYVVNYPKD NKRKMDETDA SSAVKVKRAV QKTSDLIVLG LPWKTTEQDL KEYFSTFGEV LMVQVKKDLK TGHSKGFGFV RFTEYETQVK VMSQRHMIDG RWCDCKLPNS KQSQDEPLRS RKVFVGRCTE DMTEDELREF FSQYGDVMDV FIPKPFRAFA FVTFADDQIA QSLCGEDLII KGISVHISNA EPKHNSNRQL ERSGRFGGNP GGFGNQGGFG NSRGGGAGLG NNQGSNMGGG MNFGAFSINP AMMAAAQAAL QSSWGMMGML ASQQNQSGPS GNNQNQGNMQ REPNQAFGSG NNSYSGSNSG AAIGWGSASN AGSGSG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers