missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TATA box-binding protein-like protein 1
A DNA sequence encoding the Homo sapiens (Human) TATA box-binding protein-like protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6765-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001240605.1 | |
TATA box-binding protein-like protein 1 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
9519 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
TBPL1 | |
Recombinant Protein | |
E. coli | |
MDADSDVALD ILITNVVCVF RTRCHLNLRK IALEGANVIY KRDVGKVLMK LRKPRITATI WSSGKIICTG ATSEEEAKFG ARRLARSLQK LGFQVIFTDF KVVNVLAVCN MPFEIRLPEF TKNNRPHASY EPELHPAVCY RIKSLRATLQ IFSTGSITVT GPNVKAVATA VEQIYPFVFE SRKEI |