missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tax1-binding protein 1

A DNA sequence encoding the Homo sapiens (Human) Tax1-binding protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8476-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064821

  • 5980.00 SEK / 100µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001191627.1
Tax1-binding protein 1
Research Use Only
TAX1BP3
Recombinant Protein
E. coli
MSYIPGQPVT AVVQRVEIHK LRQGENLILG FSIGGGIDQD PSQNPFSEDK TDKGIYVTRV SEGGPAEIAG LQIGDKIMQV NGWDMTMVTH DQARKRLTKR SEEVVRLLVT RQSLQKAVQQ SMLS
Greater than 90% as determined by SDS-PAGE.
30851
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers