missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tektin-5 Protein
A DNA sequence encoding the Homo sapiens (Human) Tektin-5, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_653275.1 |
---|---|
Gene ID (Entrez) | 146279 |
Name | Tektin-5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16002121
|
enQuireBio™
QP8007-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16022121
|
enQuireBio™
QP8007-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16012121
|
enQuireBio™
QP8007-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16032121
|
enQuireBio™
QP8007-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16042121
|
enQuireBio™
QP8007-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052121
|
enQuireBio™
QP8007-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_653275.1 | |
Tektin-5 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
146279 | |
Research Use Only | |
TEKT5 | |
Recombinant Protein | |
E. coli | |
MEFLGTTQTA SYCGPKKCCG LTSLPAVQAP VIQECYQPYY LPGYRYLNSW RPSLFYKIAN VQTCPDESTS TLRPPTILPT LRSALFSRYS PHDWDQSNQL QVRGAEASRL WASRLTDDSM RLLQDKDQLT HQMQEGTCRN LGQRLSDIGF WKSELSYELD RLLTENQNLE TVKRRLECAA NEVNCPLQVA LECLYHREKR IGIDLVHDNV EKNLIREVDL LKCCQEQMRK LAQRIDIQMR DNRDAQHVLE RDLEDKSSAQ CIDEKCFNLR NTSDCISFFH GMEKIDGTIS VPETWAKFSN DNIKHSQNMR ANSIQLREEA EHLFETLSDQ MWRQFTDTNL AFNARISEVT DVKNKLQTQL AKTLQEIFQA ENTIMLLERS IMAKEGPLKV AQTRLECRTR RPNMELCRDI PQLKLVNEVF TIDDTLQTLK LRLRETQDTL QLLVMTKCRL EHELAIKANT LCIDKEKCMG MRKTFPCTPR LVGHT | |
Greater than 90% as determined by SDS-PAGE. |