missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tenascin C Protein

A DNA sequence encoding the Homo sapiens (Human) Tenascin C, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8717-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016111

  • 8225.00 SEK / 200µg
Estimated Shipment: 06-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002151.2
Tenascin C Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
3371
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TNC
Recombinant Protein
E. coli
DSPRDLTATE VQSETALLTW RPPRASVTGY LLVYESVDGT VKEVIVGPDT TSYSLADLSP STHYTAKIQA LNGPLRSNMI QTIFTTIGLL YPFPKDCSQA MLNGDTTSGL YTIYLNGDKA EALEVFCDMT SDGGGWIVFL RRKNGRENFY QNWKAYAAGF GDRREEFWLG LDNLNKITAQ GQYELRVDLR DHGETAFAVY DKFSVGDAKT RYKLKVEGYS GTAGDSMAYH NGRSFSTFDK DTDSAITNCA LSYKGAFWYR NCHRVNLMGR YGDNNHSQGV NWFHWKGHEH SIQFAEMKLR PSNFRNLEGR RKRA
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers