missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Testis-expressed sequence 12 Protein
A DNA sequence encoding the Homo sapiens (Human) Testis-expressed sequence 12 protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6773-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_112565.1 | |
Testis-expressed sequence 12 Protein | |
Research Use Only | |
TEX12 | |
Recombinant Protein | |
E. coli | |
MMANHLVKPD NRNCKRPREL ESPVPDSPQL SSLGKSDSSF SEISGLFYKD EALEKDLNDV SKEINLMLST YAKLLSERAA VDASYIDEID ELFKEANAIE NFLIQKREFL RQRFTVIANT LHR | |
Greater than 90% as determined by SDS-PAGE. |
56158 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |