missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Thiamin pyrophosphokinase 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Thiamin pyrophosphokinase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6827-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16066110

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001035947.1
Thiamin pyrophosphokinase 1 Protein
Research Use Only
TPK1
Recombinant Protein
E. coli
MEHAFTPLEP LLSTGNLKYC LVILNQPLDN YFRHLWNKAL LRACADGGAN RLYDITEGER ESFLPEFING DFDSIRPEVR EYYATKGCEL ISTPDQDHTD FTKCLKMLQK KIEEKDLKVD VIVTLGGLAG RFDQIMASVN TLFQATHITP FPIIIIQEES LIYLLQPGKH RLHVDTGMEG DWCGLIPVGQ PCMQVTTTGL KWNLTNDVLA FGTLVSTSNT YDGSGVVTVE TDHPLLWTMA IKS
Greater than 80% as determined by SDS-PAGE.
27010
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers