missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thiopurine S-methyltransferase Protein
A DNA sequence encoding the Homo sapiens (Human) Thiopurine S-methyltransferase, was expressed in the hosts and tags indicated.
1580.00 SEK - 19050.00 SEK
Specifications
Accession Number | NP_000358.1 |
---|---|
Gene ID (Entrez) | 7172 |
Name | Thiopurine S-methyltransferase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16064021
|
enQuireBio™
QP8325-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16004031
|
enQuireBio™
QP8325-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054021
|
enQuireBio™
QP8325-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084021
|
enQuireBio™
QP8325-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094021
|
enQuireBio™
QP8325-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074021
|
enQuireBio™
QP8325-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000358.1 | |
Thiopurine S-methyltransferase Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
7172 | |
Research Use Only | |
TPMT | |
Recombinant Protein | |
E. coli | |
TRTSLDIEEY SDTEVQKNQV LTLEEWQDKW VNGKTAFHQE QGHQLLKKHL DTFLKGKSGL RVFFPLCGKA VEMKWFADRG HSVVGVEISE LGIQEFFTEQ NLSYSEEPIT EIPGTKVFKS SSGNISLYCC SIFDLPRTNI GKFDMIWDRG ALVAINPGDR KCYADTMFSL LGKKFQYLLC VLSYDPTKHP GPPFYVPHAE IERLFGKICN IRCLEKVDAF EERHKSWGID CLFEKLYLLT E | |
Greater than 90% as determined by SDS-PAGE. |