missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thioredoxin-2 / TXN2 Protein
A DNA sequence encoding the Homo sapiens (Human) Thioredoxin-2 / TXN2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8009-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036605.2 | |
Thioredoxin-2 / TXN2 Protein | |
Research Use Only | |
TXN2 | |
Recombinant Protein | |
E. coli | |
TTFNIQDGPD FQDRVVNSET PVVVDFHAQW CGPCKILGPR LEKMVAKQHG KVVMAKVDID DHTDLAIEYE VSAVPTVLAM KNGDVVDKFV GIKDEDQLEA FLKKLIG | |
Greater than 80% as determined by SDS-PAGE. |
25828 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |