missing translation for 'onlineSavingsMsg'
Läs mer

R&D Systems™ Recombinant Human Thrombopoietin His (HEK-293) Protein

Produktkod. 18786175
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
10 μg
100 μg
50 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18786175 1 mg
18745725 500 μg
18706175 100 μg
18765775 50 μg
18735885 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18786175 Leverantör R&D Systems™ Leverantörsnummer 288TPH01M
 Begränsad artikel
Beräknad leverans: 03-12-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Analyzed by SEC-MALS. His-tag (HEK-293-expressed)

Measured in a cell proliferation assay using MO7e human megakaryocytic leukemic cells. Avanzi, G. et al. (1988) Br. J. Haematol. 69:359. The ED50 for this effect is 0.0500-0.500 ng/mL.
TRUSTED_SUSTAINABILITY

Specifikationer

Tillträdesnummer P40225.1
För användning med (applikation) Bioactivity
Formulering Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gen-ID (Entrez) 7066
Molekylvikt (g/mol) MolecularWeight-observed: 26-30 kDa, under reducing conditions., MolecularWeight-theroretical: 19 kDa
Kvantitet 1 mg
Källa Human embryonic kidney cell, HEK293-derived human Thrombopoietin/Tpo protein Ser22-Leu195, with a C-terminal 6-His tag
Förvaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxinkoncentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF
Konjugera Unconjugated
Rekonstitution Reconstitute the 10 μg size at 100 μg/mL in PBS. Reconstitute all other sizes (at 500 μg/mL in PBS.)
Produktlinje R&D Systems
Renhet eller kvalitetsklass >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Thrombopoietin/THPO
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.