missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thymidine kinase Protein
A DNA sequence encoding the Homo sapiens (Human) Thymidine kinase, was expressed in the hosts and tags indicated.
1580.00 SEK - 26990.00 SEK
Specifications
Accession Number | YP_009137097.1 |
---|---|
Gene ID (Entrez) | 24271467 |
Name | Thymidine kinase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16095930
|
enQuireBio™
QP6799-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16015940
|
enQuireBio™
QP6799-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16005940
|
enQuireBio™
QP6799-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16025940
|
enQuireBio™
QP6799-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16035940
|
enQuireBio™
QP6799-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045940
|
enQuireBio™
QP6799-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
YP_009137097.1 | |
Thymidine kinase Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
24271467 | |
Research Use Only | |
TK | |
Recombinant Protein | |
E. coli | |
MASYPCHQHA SAFDQAARSR GHNNRRTALR PRRQQEATEV RPEQKMPTLL RVYIDGPHGM GKTTTTQLLV ALGSRDDIVY VPEPMTYWRV LGASETIANI YTTQHRLDQG EISAGDAAVV MTSAQITMGM PYAVTDAVLA PHIGGEAGSS HAPPPALTLI FDRHPIAALL CYPAARYLMG SMTPQAVLAF VALIPPTLPG TNIVLGALPE DRHIDRLAKR QRPGERLDLA MLAAIRRVYG LLANTVRYLQ CGGSWREDWG QLSGTAVPPQ GAEPQSNAGP RPHIGDTLFT LFRAPELLAP NGDLYNVFAW ALDVLAKRLR SMHVFILDYD QSPAGCRDAL LQLTSGMVQT HVTTPGSIPT ICDLARTFAR EMGEAN | |
Greater than 90% as determined by SDS-PAGE. |